3H-Imidazo[4,5-c]pyridine 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Aza-1H-benzoimidazole, 5-Azabenzimidazole

L06930.03
L06930.03EA 87.9 EUR
L06930.03
3H-Imidazo[4,5-c]pyridine 98%
3H-Imidazo[4,5-c]pyridine

Purity: 98% CAS No.: 272-97-9 Molecular Formula: C6H5N3 Formula Weight: 119.13 Melting Point: 170-173° Beilstein Reference: 2601 MDL No.: MFCD00051808

Formel: C₆H₅N₃
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051808
CAS-Nummer: 272-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR