2-Amino-3,5-dimethylbenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Dimethylanthranilic acid

L07003.06
L07003.06EA 187 EUR
L07003.06
2-Amino-3,5-dimethylbenzoesäure 98%
2-Amino-3,5-dimethylbenzoic acid

Purity: 98% CAS No.: 14438-32-5 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 189-194° Beilstein Reference: 1101785

Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Siedepunkt: 322 °C (1013 hPa)
Schmelzpunkt: 189…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017099
CAS-Nummer: 14438-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR