Dihexylether ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Purity: 98%
CAS No.: 112-58-3
Molecular Formula: C12H26O
Formula Weight: 186.34
Flash Point: 78°(172°F)
Refractive Index: 1.4204
Odour: Mild
Melting Point: -43°
Boiling Point: 228-230°
Beilstein Reference: 1738177
MDL No.: MFCD00009525
Formel:
C₁₂H₂₆O Siedepunkt: 228…230 °C Schmelzpunkt: –43 °C Dichte: 0,793 Flammpunkt: 78 °C (172 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009525 CAS-Nummer: 112-58-3 EINECS: 203-987-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...