2-(Allylthio)nicotinsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L07090.06
L07090.06EA 50.9 EUR
L07090.06
2-(Allylthio)nicotinsäure ≥98%
2-(Allylthio)nicotinsäure

Purity: 98% CAS No.: 175135-25-8 Molecular Formula: C9H9NO2S Formula Weight: 195.24 Melting Point: 144-149° MDL No.: MFCD00051681

Formel: C₉H₉NO₂S
Molecular Weight: 195.24 g/mol
Schmelzpunkt: 144…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051681
CAS-Nummer: 175135-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR