Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate 97%

Lieferant: Thermo Scientific
Achtung

L07121.06
L07121.06EA 104 EUR
L07121.06
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate 97%
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate

Purity: 97% CAS No.: 774-07-2 Molecular Formula: C7H9N3O2S Formula Weight: 199.23 Melting Point: ca 260° dec. Beilstein Reference: 176363 MDL No.: MFCD00023227

Formel: C₇H₉N₃O₂S
Molecular Weight: 199,23 g/mol
Schmelzpunkt: ∼ 260 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00023227
CAS-Nummer: 774-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR