3-Fluor-DL-phenylalanin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH

L07297.03 L07297.06
L07297.03EA 44.2 EUR
L07297.03 L07297.06
3-Fluor-DL-phenylalanin ≥98%
3-Fluor-DL-phenylalanin

Purity: 98% CAS No.: 2629-54-1 Molecular Formula: C9H10FNO2 Formula Weight: 183.18 Melting Point: ca 245° dec. Beilstein Reference: 2939807

Formel: C₉H₁₀FNO₂
Schmelzpunkt: ∼ 245 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004273
CAS-Nummer: 2629-54-1
EINECS: 220-104-1
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR