Ethyl-2,6-dimethylbenzoat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 2,6-dimethylbenzoate
L07381.06
L07381.06EA
179
EUR
L07381.06
Ethyl-2,6-dimethylbenzoat ≥98%
Ethyl-2,6-dimethylbenzoat
Purity: 98%
CAS No.: 36596-67-5
Molecular Formula: C11H14O2
Formula Weight: 178.23
Refractive Index: 1.4980
Boiling Point: 78-80°/0.9mm
MDL No.: MFCD00051785
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,23 g/mol Siedepunkt: 78…80 °C (0,9 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051785 CAS-Nummer: 36596-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...