Methyllinoleat 95%
Lieferant: Thermo Fisher Scientific
L07450.03
L07450.06
L07450.03EA
46.8
EUR
L07450.03
L07450.06
Methyllinoleat 95%
Methyllinoleat
Purity: 95%
CAS No.: 112-63-0
Molecular Formula: C19H34O2
Formula Weight: 294.47
Storage Temperature: 2°C to 8°C
Flash Point: 200°(392°F)
Refractive Index: 1.4640
Melting Point: -35°
Boiling Point: 207-208°/11mm
Beilstein Reference: 1727614
Merck Reference: 14,6095
MDL No.: MFCD00009534
Formel:
C₁₉H₃₄O₂ Siedepunkt: 207…208 °C (11 mmHg) Schmelzpunkt: –35 °C Dichte: 0,888 Flammpunkt: 200 °C (392 °F) |
MDL:
MFCD00009534 CAS-Nummer: 112-63-0 EINECS: 203-993-0 Merck Index: 14,6095 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...