Ethyl-1-benzyl-3-oxo-4-piperidincarboxylathydrochlorid 90%, tech.

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 1-Benzyl-3-oxo-4-piperidinecarboxylate HCl, Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, Ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride

L07472.03
L07472.03EA 25.9 EUR
L07472.03
Ethyl-1-benzyl-3-oxo-4-piperidincarboxylathydrochlorid 90%, tech.
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride

Purity: 90% CAS No.: 52763-21-0 Molecular Formula: C15H19NO3.HCl Formula Weight: 297.78 Melting Point: ca 158° dec. Beilstein Reference: 3749159 MDL No.: MFCD00012792

Formel: C₁₅H₂₀ClNO₃
Molecular Weight: 297,78 g/mol
Schmelzpunkt: ∼ 158 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012792
CAS-Nummer: 52763-21-0
EINECS: 258-162-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR