4-Amino-6-hydroxy-2-methylpyrimidine dihydrate ≥98%

Lieferant: Thermo Fisher Scientific

L07496.06 L07496.14
L07496.06EA 31.2 EUR
L07496.06 L07496.14
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate ≥98%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate

Purity: 98+% CAS No.: 388582-41-0 Molecular Formula: C5H7N3O.2H2O Formula Weight: 161.16 (125.13anhy) Melting Point: 300° Beilstein Reference: 118273 MDL No.: MFCD00150311

Formel: C₅H₇N₃O·2H₂O
Molecular Weight: 161.16 g/mol (125.13 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150311
CAS-Nummer: 388582-41-0
EINECS: 212-182-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR