Ethyl acetamidocyanoacetate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ethyl 2-acetamido-2-cyanoacetate

L07674.09
L07674.09EA 67.1 EUR
L07674.09
Ethyl acetamidocyanoacetate 98%
Ethyl acetamidocyanoacetate

Purity: 98% CAS No.: 4977-62-2 Molecular Formula: C7H10N2O3 Formula Weight: 170.17 Melting Point: 128-130° Beilstein Reference: 879845 MDL No.: MFCD00009133

Formel: C₇H₁₀N₂O₃
Molecular Weight: 170.168 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009133
CAS-Nummer: 4977-62-2
EINECS: 225-624-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR