ω-Brom-2-methoxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L07696.03 L07696.06
L07696.03EA 31.1 EUR
L07696.03 L07696.06
ω-Brom-2-methoxyacetophenon ≥98%
ω-Brom-2-methoxyacetophenon

Purity: 98% CAS No.: 31949-21-0 Molecular Formula: C9H9BrO2 Formula Weight: 229.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 42-46° Boiling Point: 130°/1mm Beilstein Reference: 1938788 UN No.: 3261 MDL No.: MFCD00000196

Formel: C₉H₉BrO₂
Siedepunkt: 130 °C (1 mmHg)
Schmelzpunkt: 42…46 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00000196
CAS-Nummer: 31949-21-0
EINECS: 250-870-2
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR