4,6-Diamino-2-(methylthio)pyrimidine 97%
Lieferant: Thermo Scientific
Synonyme:
2-(Methylthio)pyrimidine-4,6-diamine
L07888.14
L07888.14EA
137
EUR
L07888.14
4,6-Diamino-2-(methylthio)pyrimidine 97%
4,6-Diamino-2-(methylthio)pyrimidine
Purity: 97%
CAS No.: 1005-39-6
Molecular Formula: C5H8N4S
Formula Weight: 156.21
Melting Point: 187-191°
Beilstein Reference: 127235
MDL No.: MFCD00023241
Formel:
C₅H₈N₄S Molecular Weight: 156,21 g/mol Schmelzpunkt: 187…191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023241 CAS-Nummer: 1005-39-6 EINECS: 213-735-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...