4-Ethylbenzonitril 98%
Lieferant: Thermo Scientific
Synonyme:
p-Ethylbenzonitrile
L07921.09
L07921.09EA
98.8
EUR
L07921.09
4-Ethylbenzonitril 98%
4-Ethylbenzonitril
Purity: 98%
CAS No.: 25309-65-3
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 97°(206°F)
Refractive Index: 1.5260
Boiling Point: 237°
Beilstein Reference: 2040912
MDL No.: MFCD00016383
Formel:
C₉H₉N Molecular Weight: 131,18 g/mol Siedepunkt: 237 °C Dichte: 0,956 Flammpunkt: 97 °C (206 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016383 CAS-Nummer: 25309-65-3 EINECS: 246-811-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...