1-Chlor-4-ethylbenzol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Chloro(ethylbenzene)

L07931.06
L07931.06EA 88.5 EUR
L07931.06
1-Chlor-4-ethylbenzol ≥97%
1-Chlor-4-ethylbenzol

Purity: 97% CAS No.: 622-98-0 Molecular Formula: C8H9Cl Formula Weight: 140.61 Flash Point: 62°(144°F) Refractive Index: 1.5175 Odour: Aromatic Boiling Point: 184-185° Beilstein Reference: 1099335 MDL No.: MFCD00040855

Formel: C₈H₉Cl
Molecular Weight: 140,61 g/mol
Siedepunkt: 184…185 °C
Dichte: 1,030
Flammpunkt: 62 °C (144 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040855
CAS-Nummer: 622-98-0
EINECS: 210-763-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR