3-(4-Hydroxyphenyl)propiononitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-(4-Hydroxyphenyl)propionitrile
L07934.03
L07934.06
L07934.03EA
42.2
EUR
L07934.03
L07934.06
3-(4-Hydroxyphenyl)propiononitril ≥98%
3-(4-Hydroxyphenyl)propiononitril
Purity: 98%
CAS No.: 17362-17-3
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 54-58°
Boiling Point: 166°/8mm
UN No.: 3439
MDL No.: MFCD00016492
Formel:
C₉H₉NO Siedepunkt: 166 °C (8 mmHg) Schmelzpunkt: 54…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016492 CAS-Nummer: 17362-17-3 EINECS: 241-393-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...