Tiglinaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 497-03-0
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: 18°(64°F)
Refractive Index: 1.4480
Odour: Fruit-like
Boiling Point: 117-119°
Beilstein Reference: 1698207
UN No.: 1989
MDL No.: MFCD00006977
Formel:
C₅H₈O Siedepunkt: 117…119 °C Dichte: 0,871 Flammpunkt: 18 °C (64 °F) |
MDL:
MFCD00006977 CAS-Nummer: 497-03-0 EINECS: 207-833-0 UN: 1989 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...