Pyren ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
L08162.14
L08162.22
L08162.14EA
34.3
EUR
L08162.14
L08162.22
Pyren ≥98%
Pyrene
Purity: 98%
CAS No.: 129-00-0
Molecular Formula: C16H10
Formula Weight: 202.26
Flash Point: 208°(406°F)
Melting Point: 148-152°
Boiling Point: 393°
Beilstein Reference: 1307225
Merck Reference: 14,7963
MDL No.: MFCD00004136
Formel:
C₁₆H₁₀ Siedepunkt: 393 °C Schmelzpunkt: 148…152 °C Dichte: 1,271 Flammpunkt: 208 °C (406 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004136 CAS-Nummer: 129-00-0 EINECS: 204-927-3 UN: 3077 ADR: 9,III Merck Index: 14,7963 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...