2-(1-Pyrrolidinyl)phenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L08178.03
L08178.06
L08178.03EA
57.4
EUR
L08178.03
L08178.06
2-(1-Pyrrolidinyl)phenol ≥98%
2-(1-Pyrrolidinyl)phenol
Purity: 98%
CAS No.: 4787-77-3
Molecular Formula: C10H13NO
Formula Weight: 163.22
Melting Point: 109-112°
Beilstein Reference: 135437
Formel:
C₁₀H₁₃NO Schmelzpunkt: 109…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051659 CAS-Nummer: 4787-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...