(Phenylsulphonyl)acetic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L08299.06
L08299.14
L08299.03
L08299.06EA
114
EUR
L08299.06
L08299.14
L08299.03
(Phenylsulphonyl)acetic acid 97%
(Phenylsulphonyl)acetic acid
Purity: 97%
CAS No.: 3959-23-7
Molecular Formula: C8H8O4S
Formula Weight: 200.22
Melting Point: 109-114°
Beilstein Reference: 2504376
MDL No.: MFCD00025042
Formel:
C₈H₈O₄S Molecular Weight: 200.21 g/mol Schmelzpunkt: 109…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025042 CAS-Nummer: 3959-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...