4'-Benzyloxy-3'-methoxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific

L08430.06
L08430.06EA 90 EUR
L08430.06
4'-Benzyloxy-3'-methoxyacetophenon ≥98%
4'-Benzyloxy-3'-methoxyacetophenon

Purity: 98% CAS No.: 1835-11-6 Molecular Formula: C16H16O3 Formula Weight: 256.30 Melting Point: 85-87° Beilstein Reference: 1885776

Formel: C₁₆H₁₆O₃
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017326
CAS-Nummer: 1835-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR