6-Benzoylhexanoic acid 94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L08431.03
L08431.03EA
50.8
EUR
L08431.03
6-Benzoylhexanoic acid 94%
6-Benzoylhexanoic acid
Purity: 94%
CAS No.: 7472-43-7
Molecular Formula: C13H16O3
Formula Weight: 220.27
Melting Point: 83-86°
Beilstein Reference: 2694641
MDL No.: MFCD00039608
Formel:
C₁₃H₁₆O₃ Schmelzpunkt: 83…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039608 CAS-Nummer: 7472-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...