N- (tert-Butoxycarbonyl) -L-Asparaginsäure 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-L-aspartic Acid, Boc-Asp-OH
L08498.03
L08498.06
L08498.03EA
23.4
EUR
L08498.03
L08498.06
N- (tert-Butoxycarbonyl) -L-Asparaginsäure 98%
N- (tert-Butoxycarbonyl) -L-Asparaginsäure
Purity: 98%
CAS No.: 13726-67-5
Molecular Formula: C9H15NO6
Formula Weight: 233.23
Storage Temperature: 2°C to 8°C
Melting Point: 115°
Beilstein Reference: 1913973
Optical Rotation: -5.5° (c=1 in methanol)
MDL No.: MFCD00037279
Formel:
C₉H₁₅NO₆ Schmelzpunkt: 115 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00037279 CAS-Nummer: 13726-67-5 EINECS: 237-294-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...