Boc-D-glutamine ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-D-Gln-OH
L08536.03
L08536.03EA
50.8
EUR
L08536.03
Boc-D-glutamine ≥98%
Boc-D-glutamine
Purity: 98+%
CAS No.: 61348-28-5
Molecular Formula: C10H18N2O5
Formula Weight: 246.26
Storage Temperature: 2°C to 8°C
Melting Point: 117-119°
Beilstein Reference: 1981311
Optical Rotation: +3.5° (c=2 in ethanol)
MDL No.: MFCD00038158
Formel:
C₁₀H₁₈N₂O₅ Schmelzpunkt: 117…119 °C |
MDL:
MFCD00038158 CAS-Nummer: 61348-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...