(R)-(-)-2-Hydroxy-2-phenylpropionsäure ≥98%
Lieferant: Thermo Scientific
L08644.03
L08644.03EA
177
EUR
L08644.03
(R)-(-)-2-Hydroxy-2-phenylpropionsäure ≥98%
(R)-(-)-2-Hydroxy-2-phenylpropionsäure
Purity: 98+%
CAS No.: 3966-30-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 115-117°
Beilstein Reference: 2936598
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Schmelzpunkt: 115…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067699 CAS-Nummer: 3966-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...