2,4,6-Tribromo-3-hydroxybenzaldehyde 98%

Lieferant: Thermo Scientific
Achtung

L08656.09
L08656.09EA 55.7 EUR
L08656.09
2,4,6-Tribromo-3-hydroxybenzaldehyde 98%
2,4,6-Tribromo-3-hydroxybenzaldehyde

Purity: 98% CAS No.: 2737-22-6 Molecular Formula: C7H3Br3O2 Formula Weight: 358.81 Melting Point: 119-122° Beilstein Reference: 2098037 MDL No.: MFCD00017320

Formel: C₇H₃Br₃O₂
Molecular Weight: 358,81 g/mol
Schmelzpunkt: 119…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017320
CAS-Nummer: 2737-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR