4-(2-Methyl-2-propanyl)-2,6-dinitrophenol ≥97%

Lieferant: Thermo Scientific
Gefahr

L08670.14
L08670.14EA 54 EUR
L08670.14
4-(2-Methyl-2-propanyl)-2,6-dinitrophenol ≥97%
4-tert-Butyl-2,6-dinitrophenol

Purity: 97% CAS No.: 4097-49-8 Molecular Formula: C10H12N2O5 Formula Weight: 240.22 Melting Point: 93-95° Beilstein Reference: 1986466 UN No.: 2811 MDL No.: MFCD00051969

Formel: C₁₀H₁₂N₂O₅
Molecular Weight: 240,22 g/mol
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051969
CAS-Nummer: 4097-49-8
EINECS: 223-856-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR