3-Methoxy-N,N-dimethylanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L08700.09
L08700.09EA 103 EUR
L08700.09
3-Methoxy-N,N-dimethylanilin ≥98%
3-Methoxy-N,N-dimethylanilin

Purity: 98% CAS No.: 15799-79-8 Molecular Formula: C9H13NO Formula Weight: 151.21 Refractive Index: 1.5590 Boiling Point: 113-114°/6mm UN No.: 2810 MDL No.: MFCD00051779

Formel: C₉H₁₃NO
Siedepunkt: 113…114 °C (6 mmHg)
Dichte: 1,031
Lagertemperatur: Raumtemperatur
MDL: MFCD00051779
CAS-Nummer: 15799-79-8
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR