5,5-Dimethyl-1,3-dioxan-2-one, tech.
Lieferant: Thermo Fisher Scientific
L08767.06
L08767.03
L08767.06EA
127
EUR
L08767.06
L08767.03
5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one
CAS No.: 3592-12-9
Molecular Formula: C6H10O3
Formula Weight: 130.14
Melting Point: 105-114°
Beilstein Reference: 109055
MDL No.: MFCD00014650
Formel:
C₆H₁₀O₃ Schmelzpunkt: 105…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014650 CAS-Nummer: 3592-12-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...