5,5-Dimethyl-1,3-dioxan-2-one, tech.

Lieferant: Thermo Fisher Scientific

L08767.06 L08767.03
L08767.06EA 127 EUR
L08767.06 L08767.03
5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one

CAS No.: 3592-12-9 Molecular Formula: C6H10O3 Formula Weight: 130.14 Melting Point: 105-114° Beilstein Reference: 109055 MDL No.: MFCD00014650

Formel: C₆H₁₀O₃
Molecular Weight: 130.14 g/mol
Schmelzpunkt: 105…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014650
CAS-Nummer: 3592-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR