Natriumbis(trimethylsilyl)amid 1 M in THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Natriumbis(trimethylsilyl)amid, HMDS Natriumsalz, N-Natriohexamethyldisilazan, HMDS-Na
L08779.AE
L08779.AP
L08779.AEEA
79.7
EUR
L08779.AE
L08779.AP
Natriumbis(trimethylsilyl)amid 1 M in THF
Natriumbis(trimethylsilyl)amid
CAS No.: 1070-89-9
Molecular Formula: C6H18NNaSi2
Formula Weight: 183.38
Flash Point: -17°(1°F)
Beilstein Reference: 3629917
UN No.: 2924
MDL No.: MFCD00009835
Formel:
C₆H₁₈NNaSi₂ Molecular Weight: 183.38 g/mol Dichte: 0.904 Flammpunkt: –17 °C (1 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009835 CAS-Nummer: 1070-89-9 EINECS: 213-983-8 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...