2-Fluor-6-nitrophenol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L08851.03 L08851.06
L08851.03EA 56.8 EUR
L08851.03 L08851.06
2-Fluor-6-nitrophenol ≥98%
2-Fluor-6-nitrophenol

Purity: 98+% CAS No.: 1526-17-6 Molecular Formula: C6H4FNO3 Formula Weight: 157.10 Melting Point: 91-94° Beilstein Reference: 1870307

Formel: C₆H₄FNO₃
Schmelzpunkt: 91…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042446
CAS-Nummer: 1526-17-6
EINECS: 216-199-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR