6-Methylnicotinsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Methyl-3-pyridincarbonsäure, 6-Methylpyridin-3-carbonsäure
L08877.03
L08877.06
L08877.14
L08877.03EA
28.6
EUR
L08877.03
L08877.06
L08877.14
6-Methylnicotinsäure ≥99%
6-Methylnicotinsäure
Purity: 99%
CAS No.: 3222-47-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 210-215°
Beilstein Reference: 112049
Formel:
C₇H₇NO₂ Schmelzpunkt: 210…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006341 CAS-Nummer: 3222-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...