Methyl-3-methoxy-2-nitrobenzoat ≥98%
Lieferant: Thermo Scientific
Synonyme:
3-Methoxy-2-nitrobenzoesäuremethylester
L08886.06
L08886.06EA
79.5
EUR
L08886.06
Methyl-3-methoxy-2-nitrobenzoat ≥98%
Methyl-3-methoxy-2-nitrobenzoat
Purity: 98%
CAS No.: 5307-17-5
Molecular Formula: C9H9NO5
Formula Weight: 211.18
Melting Point: 142-144°
Beilstein Reference: 2697098
Formel:
C₉H₉NO₅ Molecular Weight: 211,17 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051968 CAS-Nummer: 5307-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...