Ethyl-N,N-dimethyloxamate 98%
Lieferant: Thermo Fisher Scientific
L08923.14
L08923.06
L08923.14EA
63
EUR
L08923.14
L08923.06
Ethyl-N,N-dimethyloxamate 98%
Ethyl-N,N-dimethyloxamate
Purity: 98%
CAS No.: 16703-52-9
Molecular Formula: C6H11NO3
Formula Weight: 145.16
Flash Point:
Formel:
C₆H₁₁NO₃ Siedepunkt: 118…120 °C (10 mmHg) Dichte: 1,068 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015153 CAS-Nummer: 16703-52-9 EINECS: 240-755-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...