Ethyl-N,N-dimethyloxamate 98%

Lieferant: Thermo Fisher Scientific

L08923.14 L08923.06
L08923.14EA 63 EUR
L08923.14 L08923.06
Ethyl-N,N-dimethyloxamate 98%
Ethyl-N,N-dimethyloxamate

Purity: 98% CAS No.: 16703-52-9 Molecular Formula: C6H11NO3 Formula Weight: 145.16 Flash Point: 110°(230°F) Refractive Index: 1.4430 Boiling Point: 118-120°/10mm Beilstein Reference: 1906209 MDL No.: MFCD00015153

Formel: C₆H₁₁NO₃
Siedepunkt: 118…120 °C (10 mmHg)
Dichte: 1,068
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015153
CAS-Nummer: 16703-52-9
EINECS: 240-755-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR