4-Benzyl-L-aspartate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester
L08956.06
L08956.14
L08956.06EA
71.9
EUR
L08956.06
L08956.14
4-Benzyl-L-aspartate 98%
4-Benzyl-L-aspartate
Purity: 98%
CAS No.: 2177-63-1
Molecular Formula: C11H13NO4
Formula Weight: 223.23
Melting Point: ca 220° dec.
Beilstein Reference: 1983183
Optical Rotation: +27.5° (c=1 in 1N HCl)
MDL No.: MFCD00037208
Formel:
C₁₁H₁₃NO₄ Schmelzpunkt: ∼ 220 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037208 CAS-Nummer: 2177-63-1 EINECS: 218-541-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...