N-Methylphthalimid ≥98%

Lieferant: Thermo Scientific

Synonyme: N-Methylphthalsäureimid, N-Methyl-1,2-benzoldicarbonsäureimid

L08991.14
L08991.14EA 75 EUR
L08991.14
N-Methylphthalimid ≥98%
N-Methylphthalimid

Purity: 98% CAS No.: 550-44-7 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 132-136° Beilstein Reference: 124428

Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
Siedepunkt: 286 °C (1013 hPa)
Schmelzpunkt: 132…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023063
CAS-Nummer: 550-44-7
EINECS: 208-982-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR