Methyl-2-methylnicotinat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate
L09074.06
L09074.14
L09074.06EA
77.2
EUR
L09074.06
L09074.14
Methyl-2-methylnicotinat ≥97%
Methyl-2-methylnicotinat
Purity: 97%
CAS No.: 65719-09-7
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Refractive Index: 1.5165
Boiling Point: 102-104°/10mm
MDL No.: MFCD00051961
Formel:
C₈H₉NO₂ Siedepunkt: 102…104 °C (10 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051961 CAS-Nummer: 65719-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...