DL-Homocysteinthiolactonhydrochlorid 99%

Lieferant: Thermo Fisher Scientific

Synonyme: DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl

L09077.14 L09077.22
L09077.14EA 31.5 EUR
L09077.14 L09077.22
DL-Homocysteinthiolactonhydrochlorid 99%
DL-Homocysteinethiolactone hydrochloride

Purity: 99% CAS No.: 6038-19-3 Molecular Formula: C4H7NOS.HCl Formula Weight: 153.63 Storage Temperature: 2°C to 8°C Melting Point: ca 204° dec. Beilstein Reference: 3562103 MDL No.: MFCD00012724

Formel: C₄H₇NOS
Schmelzpunkt: ∼ 204 °C (Zers.)
MDL: MFCD00012724
CAS-Nummer: 6038-19-3
EINECS: 227-923-3
Merck Index: 14,4734

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR