DL-Homocysteinthiolactonhydrochlorid 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl
L09077.14
L09077.22
L09077.14EA
31.5
EUR
L09077.14
L09077.22
DL-Homocysteinthiolactonhydrochlorid 99%
DL-Homocysteinethiolactone hydrochloride
Purity: 99%
CAS No.: 6038-19-3
Molecular Formula: C4H7NOS.HCl
Formula Weight: 153.63
Storage Temperature: 2°C to 8°C
Melting Point: ca 204° dec.
Beilstein Reference: 3562103
MDL No.: MFCD00012724
Formel:
C₄H₇NOS Schmelzpunkt: ∼ 204 °C (Zers.) |
MDL:
MFCD00012724 CAS-Nummer: 6038-19-3 EINECS: 227-923-3 Merck Index: 14,4734 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...