Methylthymolblau Natriumsalz
Lieferant: Thermo Scientific
Synonyme:
Methylthymolblau-Komplexon, Methylthymolblau Tetranatriumsalz, Methyl thymol blue complexone
L09381.06
L09381.06EA
69.4
EUR
L09381.06
Methylthymolblau Natriumsalz
Methylthymolblau Natriumsalz
CAS No.: 1945-77-3
Molecular Formula: C37H40N2Na4O13S
Formula Weight: 844.76
MDL No.: MFCD00148905
Formel:
C₃₇H₄₀N₂Na₄O₁₃S Molecular Weight: 844,76 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148905 CAS-Nummer: 1945-77-3 EINECS: 217-743-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...