cis-4-Cyclohexen-1,2-dicarbonsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L09392.14
L09392.22
L09392.14EA
38.1
EUR
L09392.14
L09392.22
cis-4-Cyclohexen-1,2-dicarbonsäure ≥98%
cis-4-Cyclohexen-1,2-dicarbonsäure
Purity: 98%
CAS No.: 2305-26-2
Molecular Formula: C8H10O4
Formula Weight: 170.17
Melting Point: 165-170°
Beilstein Reference: 2050419
Formel:
C₈H₁₀O₄ Schmelzpunkt: 165…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070482 CAS-Nummer: 2305-26-2 EINECS: 218-974-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...