Tetraethylharnstoff ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,1,3,3-Tetraethylurea

L09458.06 L09458.14
L09458.06EA 28.8 EUR
L09458.06 L09458.14
Tetraethylharnstoff ≥99%
Tetraethylharnstoff

Purity: 99% CAS No.: 1187-03-7 Molecular Formula: (CH3CH2)2NCON(CH2CH3)2 Formula Weight: 172.27 Flash Point: 81°(177°F) Refractive Index: 1.4470 Boiling Point: 212-214° Beilstein Reference: 1763126 MDL No.: MFCD00042881

Formel: C₉H₂₀N₂O
Molecular Weight: 172.27 g/mol
Siedepunkt: 212…214 °C
Dichte: 0.910
Flammpunkt: 81 °C (177 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042881
CAS-Nummer: 1187-03-7
EINECS: 214-696-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR