(2-Bromethyl)cyclohexan ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L09466.14 L09466.06 L09466.03
L09466.14EA 264 EUR
L09466.14 L09466.06 L09466.03
(2-Bromethyl)cyclohexan ≥99%
(2-Bromethyl)cyclohexan

Purity: 99% CAS No.: 1647-26-3 Molecular Formula: C8H15Br Formula Weight: 191.12 Flash Point: 96°(204°F) Refractive Index: 1.4900 Boiling Point: 70-71°/6mm Beilstein Reference: 1919816 MDL No.: MFCD00019398

Formel: C₈H₁₅Br
Siedepunkt: 70…71 °C (6 mmHg)
Dichte: 1,221
Flammpunkt: 96 °C (204 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00019398
CAS-Nummer: 1647-26-3
EINECS: 216-712-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR