DL-2,3-Diaminopropionic acid hydrochloride 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,3-Diaminopropanoic acid hydrochloride, 3-Amino-DL-alanine Hydrochloride, DL-2,3-Diaminopropionic acid monohydrochloride

L09485.06
L09485.06EA 118 EUR
L09485.06
DL-2,3-Diaminopropionic acid hydrochloride 99%
DL-2,3-Diaminopropionic acid hydrochloride

Purity: 99% CAS No.: 54897-59-5 Molecular Formula: C3H8N2O2.HCl Formula Weight: 140.57 Melting Point: ca 232° dec. Beilstein Reference: 5293359 MDL No.: MFCD00012884

Formel: C₃H₈N₂O₂
Molecular Weight: 140.57 g/mol
Schmelzpunkt: ∼ 232 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012884
CAS-Nummer: 54897-59-5
EINECS: 259-387-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR