3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Hydroxy-2H,2H-perfluoro-3-methylbutanoic acid
L09516.06
L09516.06EA
170
EUR
L09516.06
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid 97%
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid
Purity: 97%
CAS No.: 1547-36-0
Molecular Formula: C5H4F6O3
Formula Weight: 226.08
Melting Point: 85-89°
Beilstein Reference: 1911706
UN No.: UN3261
MDL No.: MFCD00040910
Formel:
C₅H₄F₆O₃ Schmelzpunkt: 85…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040910 CAS-Nummer: 1547-36-0 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...