(1R,2R)-(+)-1,2-Diphenylethylendiamin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L09558.03
L09558.MD
L09558.03EA
58.8
EUR
L09558.03
L09558.MD
(1R,2R)-(+)-1,2-Diphenylethylendiamin ≥98%
(1R,2R)-(+)-1,2-Diphenylethylendiamin
Purity: 98+%
CAS No.: 35132-20-8
Molecular Formula: C14H16N2
Formula Weight: 212.30
Storage Temperature: 2°C to 8°C
Melting Point: 80-84°
Beilstein Reference: 2369988
Optical Rotation: +104° (c=1 in methanol)
UN No.: 3259
MDL No.: MFCD00082769
Formel:
C₁₄H₁₆N₂ Schmelzpunkt: 80…84 °C |
MDL:
MFCD00082769 CAS-Nummer: 35132-20-8 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...