(S)-(+)-2-(Methoxymethyl)pyrrolidin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L09608.03 L09608.06
L09608.03EA 74.3 EUR
L09608.03 L09608.06
(S)-(+)-2-(Methoxymethyl)pyrrolidin ≥98%
(S)-(+)-2-(Methoxymethyl)pyrrolidin

Purity: 98% CAS No.: 63126-47-6 Molecular Formula: C6H13NO Formula Weight: 115.18 Flash Point: 45°(113°F) Refractive Index: 1.4457 Boiling Point: 61-62°/40mm Beilstein Reference: 3587324 Optical Rotation: +2.2° (c=2 in benzene) UN No.: 1993 MDL No.: MFCD00010408

Formel: C₆H₁₃NO
Siedepunkt: 61…62 °C (40 mmHg)
Dichte: 0,933
Flammpunkt: 45 °C (113 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010408
CAS-Nummer: 63126-47-6
EINECS: 626-212-9
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR