L-Azetidin-2-Carboxylsäure 98%

Lieferant: Thermo Scientific

Synonyme: L-Trimethyleneimine-2-carboxylic Acid, (S)-Azetidine-2-carboxylic acid, (S)-(-)-2-Azetidinecarboxylic acid, L-Azetidine-2-carboxylic Acid [Antagonist of L-Proline]

L09668.MD
L09668.MDEA 120 EUR
L09668.MD
L-Azetidin-2-Carboxylsäure 98%
L-Azetidine-2-carboxylic acid

Purity: 99% CAS No.: 2133-34-8 Molecular Formula: C4H7NO2 Formula Weight: 101.11 Melting Point: ca 217° dec. Beilstein Reference: 80678 Optical Rotation: -123° (c=4 in H2O) MDL No.: MFCD00005166

Formel: C₄H₇NO₂
Molecular Weight: 101,11 g/mol
Schmelzpunkt: ∼ 217 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005166
CAS-Nummer: 2133-34-8
EINECS: 218-362-5
Merck Index: 14,908

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR